Merck
  • A single cDNA, hTFIIA/alpha, encodes both the p35 and p19 subunits of human TFIIA.
A single cDNA, hTFIIA/alpha, encodes both the p35 and p19 subunits of human TFIIA.
Genes & development (1993-11-01)
J DeJong, R G Roeder
ABSTRACT
TFIIA is a transcription factor that, by interacting with the TATA-binding subunit (TBP) of TFIID, modulates transcription initiation by RNA polymerase II in vitro. By use of a mobility shift assay, TFIIA was purified from HeLa cells as a complex of 35-, 19-, and 12-kD subunits. Oligonucleotides were used to isolate a human cDNA clone, hTFIIA/alpha, which encodes a 55-kD protein with homology to the product of the yeast gene TOA1. The open reading frame of hTFIIA/alpha contains peptide sequences obtained from both the p35 and p19 subunits of natural human TFIIA, and thus encodes these two subunits. Consistent with this, antiserum raised against the 55-kD hTFIIA/alpha-encoded protein reacted with both the p35 and p19 subunits of natural TFIIA, and the recombinant protein could functionally replace those subunits in a mobility shift assay with renatured p12. An efficient affinity purification for natural human TFIIA was suggested by the sequence of the hTFIIA/alpha protein and demonstrated biochemically. Finally, transcription from the adenovirus major late promoter was greatly reduced in nuclear extracts depleted with anti-TFIIA/alpha serum and was restored to original levels by the readdition of purified human TFIIA.
Sigma-Aldrich
TFIIA, p55 subunit, GST tagged human, recombinant, expressed in E. coli, ≥70% (SDS-PAGE)
Sigma-Aldrich
TFIIA-p55 human, recombinant, expressed in E. coli, ≥80% (SDS-PAGE)
Sigma-Aldrich
TFIIA, reconstituted human, recombinant, expressed in E. coli, ≥80% (SDS-PAGE)