Merck
  • [Determination of 92 pesticide residues in tea by gas chromatography with solid-phase extraction].
[Determination of 92 pesticide residues in tea by gas chromatography with solid-phase extraction].
Se pu = Chinese journal of chromatography (2009-01-24)
Zhengyun Lou, Zongmao Chen, Fengjian Luo, Fubin Tang, Guangming Liu
ABSTRACT
A multiresidue analytical method was developed for the determination of ninety-two pesticides in tea using solid-phase extraction (SPE) and gas chromatographic determination. Tea samples were extracted with acetonitrile, organophosphorus pesticides were clean-up by an Envi-Carb SPE cartridge, eluted with 10 mL acetonitrile-toluene (3:1, v/v) and determined by gas chromatography-flame photometric detection (GC-FPD); organochlorine pesticides and pyrethroids pesticides were cleaned-up by Envi-Carb + NH2 SPE cartridges, eluted with 5 mL acetonitrile-toluene (3:1, v/v) and determined by gas chromatography-electron capture detection (GC-ECD). The recoveries of those pesticides were ranged from 80.3% to 117.1% by fortification test with the relative standard deviations being from 1.5% to 9.8%. The limits of detection were 0.0025 - 0.10 mg/kg. The proposed method is characterized by simplicity, higher sensitivity and accuracy.
Product Description
Supelco
Supel QuE, PSA/C18/ENVI-Carb Tube, pack of 100, AOAC® 2007.01
Supelco
Supel QuE, PSA/ENVI-Carb Tube 1, pk of 50, suitable for EN 15662:2008 per BS, centrifuge tube volume 15 mL , Shaker Compatible
Supelco
Supel QuE, PSA/ENVI-Carb Tube 2, pk of 50, suitable for EN 15662:2008 per BS, centrifuge tube volume 15 mL , Shaker Compatible
Supelco
Supel QuE, PSA/C18/ENVI-Carb Tube, pk of 50, AOAC® 2007.01, centrifuge tube volume 15 mL , Shaker Compatible
Supelco
Supel QuE, PSA/ENVI-Carb (EN) Cleanup Tube 1, pack of 100, suitable for EN 15662:2008 per BS
Supelco
Supel QuE, PSA/ENVI-Carb (EN) Cleanup Tube 2, pack of 100, suitable for EN 15662:2008 per BS