Skip to Content
Merck
vibrant-m

Zolmitriptan Related Compound B

vibrant-m
Synonym(s):
(S)-β-Amino-3-[2-(dimethylamino)ethyl]-1H-indole-5-propanol, (S)-2-Amino-3-{3-[2-(dimethylamino)ethyl]-1H-indol-5-yl}propan-1-ol
Empirical Formula (Hill Notation):
C15H23N3O
CAS Number:
Molecular Weight:
261.36

Products

Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1