Skip to Content
Merck
Zirconium(IV) chloride ≥99.9% trace metals basis

Zirconium(IV) chloride

Zirconium(IV) chloride ≥99.9% trace metals basis
Synonym(s):
Tetrachlorozirconium, Zirconium tetrachloride, Zirconium(IV) chloride
Linear Formula:
ZrCl4
CAS Number:
Molecular Weight:
233.04
EC Number:

Products

Brand
Sustainability
Feature
Formula Weight
Physical Form
Purity
Quality Segment
Application
Reagent Type
Core
Special Grade
Showing 1-4 of 4