![Vinylbenzyl chloride purum, mixture of isomers, ≥95.0%, stabilized](/deepweb/assets/sigmaaldrich/product/structures/137/974/a858a46c-7d35-47a0-80f6-7c811822ccde/640/a858a46c-7d35-47a0-80f6-7c811822ccde.png)
Vinylbenzyl chloride
![Vinylbenzyl chloride purum, mixture of isomers, ≥95.0%, stabilized](/deepweb/assets/sigmaaldrich/product/structures/137/974/a858a46c-7d35-47a0-80f6-7c811822ccde/640/a858a46c-7d35-47a0-80f6-7c811822ccde.png)
Synonym(s):
(Chloromethyl)styrene
Linear Formula:
H2C=CHC6H4CH2Cl
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Monomer Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing