![Vanillic acid 4-β-D-glucoside phyproof® Reference Substance](/deepweb/assets/sigmaaldrich/product/structures/160/813/da46400d-4d27-4d40-94d6-6b0c7470a5cd/640/da46400d-4d27-4d40-94d6-6b0c7470a5cd.png)
Vanillic acid 4-β-D-glucoside
![Vanillic acid 4-β-D-glucoside phyproof® Reference Substance](/deepweb/assets/sigmaaldrich/product/structures/160/813/da46400d-4d27-4d40-94d6-6b0c7470a5cd/640/da46400d-4d27-4d40-94d6-6b0c7470a5cd.png)
Synonym(s):
4-Hydroxy-3-methoxybenzoic acid 4-β-D-glucoside, Vanillic acid 4-β-D-glucopyranoside
Empirical Formula (Hill Notation):
C14H18O9
CAS Number:
Molecular Weight:
330.29
Products
Formula Weight
Manufacturer
Physical Form
Product Line
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing