Tylosin Tartrate
Synonym(s):
Tylosin (2R,3R)-2, 3-dihydroxybutanedioate (salt)
Linear Formula:
(C46H77NO17)x · C4H6O6
CAS Number:
Products
Brand
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique