
Trinexapac-ethyl

Empirical Formula (Hill Notation):
C13H16O5
CAS Number:
Molecular Weight:
252.26
Products
Product Category
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2