Skip to Content
Merck
Trimethylboroxine 50% THF solution

Trimethylboroxine

Trimethylboroxine 50% THF solution
Empirical Formula (Hill Notation):
C3H9B3O3
CAS Number:
Molecular Weight:
125.53

Products

Shipping
Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Showing 1-2 of 2
Compare
Product Number
Description
Pricing