Skip to Content
Merck
trans-Anethole Pharmaceutical Secondary Standard; Certified Reference Material

trans-Anethole

trans-Anethole Pharmaceutical Secondary Standard; Certified Reference Material
Synonym(s):
4-Propenylanisole, trans-1-Methoxy-4-(1-propenyl)benzene, trans-Anethole
Linear Formula:
CH3CH=CHC6H4OCH3
CAS Number:
Molecular Weight:
148.20
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
API Family
Reference Material Type
Special Grade
Technique