Skip to Content
Merck
Tolmetin sodium dihydrate ≥98% (HPLC)

Tolmetin sodium dihydrate

Tolmetin sodium dihydrate ≥98% (HPLC)
Synonym(s):
Tolmetin sodium salt dihydrate, 1-Methyl-5-(p-toluoyl)pyrrole-2-acetic acid sodium salt dihydrate
Linear Formula:
C15H14NO3Na · 2H2O
CAS Number:
Molecular Weight:
315.30

Products

Product Category
Brand
Color
Formula Weight
Gene Alias
Physical Form
Purity
Research Area
Showing 1-1 of 1
Compare
Product Number
Description
Pricing