Skip to Content
Merck
Titanyl phthalocyanine Dye content 95 %

Titanyl phthalocyanine

Titanyl phthalocyanine Dye content 95 %
Empirical Formula (Hill Notation):
C32H16N8OTi
CAS Number:
Molecular Weight:
576.39

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Chemical Composition
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing