![Thifluzamide PESTANAL®, analytical standard](/deepweb/assets/sigmaaldrich/product/structures/389/399/914dd074-7b96-4f39-8867-7a82066435d2/640/914dd074-7b96-4f39-8867-7a82066435d2.png)
Thifluzamide
![Thifluzamide PESTANAL®, analytical standard](/deepweb/assets/sigmaaldrich/product/structures/389/399/914dd074-7b96-4f39-8867-7a82066435d2/640/914dd074-7b96-4f39-8867-7a82066435d2.png)
Synonym(s):
2′,6′-Dibromo-2-methyl-4′-trifluoromethoxy-4-trifluoromethyl-1,3-thiazole-5-carboxanilide, N-[2,6-Dibromo-4-(trifluoromethoxy)phenyl]-2-methyl-4-(trifluoromethyl)-5-thiazolecarboxamide
Empirical Formula (Hill Notation):
C13H6Br2F6N2O2S
CAS Number:
Molecular Weight:
528.06
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Purity
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2