Skip to Content
Merck
THC-O-Acetate solution 1 mg/mL in acetonitrile, certified reference material, ampule of 1 mL, Cerilliant®

THC-O-Acetate solution

THC-O-Acetate solution 1 mg/mL in acetonitrile, certified reference material, ampule of 1 mL, Cerilliant®
Synonym(s):
THC-O-Acetate solution
Empirical Formula (Hill Notation):
C23H32O3
CAS Number:
Molecular Weight:
356.50

Products

Brand
Formula Weight
Manufacturer
Format
Quality Segment
Package Size
Reference Material Type
Special Grade