
Temazepam Related Compound A

Synonym(s):
5-Chloro-2-(methylamino)benzophenone
Linear Formula:
CH3NHC6H3(Cl)COC6H5
CAS Number:
Molecular Weight:
245.70
Products
Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing