![Tebutam PESTANAL®, analytical standard](/deepweb/assets/sigmaaldrich/product/structures/126/832/d53bc793-c430-4bcb-9aa9-8d14642c60e8/640/d53bc793-c430-4bcb-9aa9-8d14642c60e8.png)
Tebutam
![Tebutam PESTANAL®, analytical standard](/deepweb/assets/sigmaaldrich/product/structures/126/832/d53bc793-c430-4bcb-9aa9-8d14642c60e8/640/d53bc793-c430-4bcb-9aa9-8d14642c60e8.png)
Synonym(s):
N-Benzyl-N-isopropylpivalamide
Empirical Formula (Hill Notation):
C15H23NO
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2