
Tebbe reagent solution

Synonym(s):
Bis(cyclopentadienyl)-μ-chloro(dimethylaluminum)-μ-methylenetitanium
Empirical Formula (Hill Notation):
C13H18AlClTi
CAS Number:
Molecular Weight:
284.58
Products
Formula Weight
Physical Form
Reaction Type
Showing 1-1 of 1