Skip to Content
Merck
Sodium pyruvate-3-13C endotoxin tested, 99 atom % 13C, 98% (CP)

Sodium pyruvate-3-13C

Sodium pyruvate-3-13C endotoxin tested, 99 atom % 13C, 98% (CP)
Synonym(s):
Pyruvic acid-3-13C sodium salt
Linear Formula:
13CH3COCO2Na
CAS Number:
Molecular Weight:
111.04

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Isotope
Markush Class
Markush Group
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing