![Sodium acetate-1-13C API for Clinical Studies, 99 atom % 13C](/deepweb/assets/sigmaaldrich/product/structures/295/961/28360aee-fa52-4359-b1c8-07e211954173/640/28360aee-fa52-4359-b1c8-07e211954173.png)
Sodium acetate-1-13C
![Sodium acetate-1-13C API for Clinical Studies, 99 atom % 13C](/deepweb/assets/sigmaaldrich/product/structures/295/961/28360aee-fa52-4359-b1c8-07e211954173/640/28360aee-fa52-4359-b1c8-07e211954173.png)
Synonym(s):
Acetic acid-1-13C sodium salt
Linear Formula:
CH3 13CO2Na
CAS Number:
Molecular Weight:
83.03
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Quality Segment
Isotope
Markush Class
Markush Group
Special Grade
Technique
Showing 1-3 of 3