![Silylating mixture II according to Horning for GC derivatization, LiChropur™](/deepweb/assets/sigmaaldrich/product/images/180/925/c78867db-c35d-4d7b-b463-35f28d6f859c/640/c78867db-c35d-4d7b-b463-35f28d6f859c.jpg)
Silylating mixture II according to Horning
![Silylating mixture II according to Horning for GC derivatization, LiChropur™](/deepweb/assets/sigmaaldrich/product/images/180/925/c78867db-c35d-4d7b-b463-35f28d6f859c/640/c78867db-c35d-4d7b-b463-35f28d6f859c.jpg)
Synonym(s):
N,O-Bis(trimethylsilyl)acetamide – chlorotrimethylsilane – 1-(trimethylsilyl)imidazole mixture
CAS Number:
Products
Brand
Physical Form
Quality Segment
Reaction Type
Reagent Type
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing