![Silanization solution I ~5% (dimethyldichlorosilane in heptane), Selectophore™](/deepweb/assets/sigmaaldrich/product/structures/416/064/38579887-db1c-4429-8cd5-1dbcb86ff72e/640/38579887-db1c-4429-8cd5-1dbcb86ff72e.png)
Silanization solution I
![Silanization solution I ~5% (dimethyldichlorosilane in heptane), Selectophore™](/deepweb/assets/sigmaaldrich/product/structures/416/064/38579887-db1c-4429-8cd5-1dbcb86ff72e/640/38579887-db1c-4429-8cd5-1dbcb86ff72e.png)
Synonym(s):
Dichlorodimethylsilane solution, Dimethyldichlorosilane solution
Empirical Formula (Hill Notation):
C2H6Cl2Si
CAS Number:
Molecular Weight:
129.06
Products
Brand
Formula Weight
Physical Form
Product Line
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing