![Sertraline for peak identification European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/114/205/bff38abc-3789-4ca3-b75a-525529428fd6/640/bff38abc-3789-4ca3-b75a-525529428fd6.png)
Sertraline for peak identification
![Sertraline for peak identification European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/114/205/bff38abc-3789-4ca3-b75a-525529428fd6/640/bff38abc-3789-4ca3-b75a-525529428fd6.png)
Synonym(s):
Sertraline hydrochloride, (1S,4S)-4-(3,4-Dichlorophenyl)-1,2,3,4-tetrahydro-N-methyl-1-naphthalenamine hydrochloride
Empirical Formula (Hill Notation):
C17H17NCl2 · HCl
CAS Number:
Molecular Weight:
342.69
Products
Formula Weight
Gene Alias
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing