(S)-(+)-1-Aminoindan
Synonym(s):
(S)-(+)-1-Indanamine
Empirical Formula (Hill Notation):
C9H11N
CAS Number:
Molecular Weight:
133.19
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Gene Alias
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing