![(S)-(−)-Bay K8644 ≥98% (HPLC), solid](/deepweb/assets/sigmaaldrich/product/structures/422/947/a8d051dc-1cf7-446f-a408-3211b373ef64/640/a8d051dc-1cf7-446f-a408-3211b373ef64.png)
(S)-(−)-Bay K8644
![(S)-(−)-Bay K8644 ≥98% (HPLC), solid](/deepweb/assets/sigmaaldrich/product/structures/422/947/a8d051dc-1cf7-446f-a408-3211b373ef64/640/a8d051dc-1cf7-446f-a408-3211b373ef64.png)
Synonym(s):
S-(−)-1,4-Dihydro-2,6-dimethyl-5-nitro-4-[2-(trifluoromethyl)phenyl]-3-pyridinecarboxylic acid methyl ester
Empirical Formula (Hill Notation):
C16H15F3N2O4
CAS Number:
Molecular Weight:
356.30
Products
Product Category
Brand
Color
Formula Weight
Gene Alias
Physical Form
Purity
Research Area
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing