Skip to Content
Merck
Ristomycin monosulfate ≥90% ristocetin A basis (balance primarily Ristocetin B)

Ristomycin monosulfate

Ristomycin monosulfate ≥90% ristocetin A basis (balance primarily Ristocetin B)
Synonym(s):
Ristocetin sulfate salt
Empirical Formula (Hill Notation):
C95H110N8O44 · H2SO4
CAS Number:
Molecular Weight:
2166.00

Products

Product Category
Brand
Formula Weight
Purity
Research Area
Mode of Action