
Repaglinide Related Compound C

Synonym(s):
(S)-2-Ethoxy-4-[2-oxo -2-[[2-phenyl-1-[2-(1-piperidinyl)phenyl]ethyl]amino]ethyl]benzoic acid
Empirical Formula (Hill Notation):
C30H34N2O4
CAS Number:
Molecular Weight:
486.60
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2