Skip to Content
Merck
(R)-α-Acryloyloxy-β,β-dimethyl-γ-butyrolactone 95%

(R)-α-Acryloyloxy-β,β-dimethyl-γ-butyrolactone

(R)-α-Acryloyloxy-β,β-dimethyl-γ-butyrolactone 95%
Empirical Formula (Hill Notation):
C9H12O4
CAS Number:
Molecular Weight:
184.19

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing