
Purmorphamine

Synonym(s):
2-(1-Naphthoxy)-6-(4-morpholinoanilino)-9-cyclohexylpurine; 9-Cyclohexyl-N-[4-(4-morpholinyl)phenyl]-2-(1-naphthalenyloxy)-, Purmorphamine
Empirical Formula (Hill Notation):
C31H32N6O2
CAS Number:
Molecular Weight:
520.62
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Showing 1-2 of 2
Compare
Product Number
Description
Pricing