![propylene glycol dilaurate AldrichCPR](/deepweb/assets/sigmaaldrich/product/structures/927/384/a626de47-6b43-43c3-b6c6-b5b3ddbdbbb6/640/a626de47-6b43-43c3-b6c6-b5b3ddbdbbb6.png)
propylene glycol dilaurate
![propylene glycol dilaurate AldrichCPR](/deepweb/assets/sigmaaldrich/product/structures/927/384/a626de47-6b43-43c3-b6c6-b5b3ddbdbbb6/640/a626de47-6b43-43c3-b6c6-b5b3ddbdbbb6.png)
Synonym(s):
Dodecanoic acid 1-methyl-1,2-ethanediyl ester
Empirical Formula (Hill Notation):
C27H52O4
CAS Number:
Molecular Weight:
440.70
Products
Product Category
Brand
Formula Weight
Manufacturer
Physical Form
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-3 of 3