Skip to Content
Merck
Promethazine hydrochloride British Pharmacopoeia (BP) Reference Standard

Promethazine hydrochloride

Promethazine hydrochloride British Pharmacopoeia (BP) Reference Standard
Synonym(s):
10-[2-(Dimethylamino)propyl]phenothiazine hydrochloride, Promethazine hydrochloride
Empirical Formula (Hill Notation):
C17H20N2S · HCl
CAS Number:
Molecular Weight:
320.88
EC Number:

Products

Product Category
Brand
Formula Weight
Gene Alias
Manufacturer
Physical Form
Agency
Format
Research Area
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique