Skip to Content
Merck
vibrant-m

Primaquine Related Compound A

vibrant-m
Synonym(s):
Quinocide, 8-(4-Aminopentylamino)-6-methoxyquinoline, N1-(6-Methoxyquinolyl)-1,4-pentanediamine
Empirical Formula (Hill Notation):
C15H21N3O
CAS Number:
Molecular Weight:
259.35

Products

Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1