
Potassium chloride solution Certified Reference Material

Synonym(s):
Potassium chloride solution
Linear Formula:
KCl
CAS Number:
Molecular Weight:
74.55
Products
Brand
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Showing 1-3 of 3
Compare
Product Number
Description
Pricing