Skip to Content
Merck
Perindopril Related Compound B certified reference material, pharmaceutical secondary standard

Perindopril Related Compound B

Perindopril Related Compound B certified reference material, pharmaceutical secondary standard
Synonym(s):
Perindoprilat, (2S,3aS,7aS)-1-[(2S)-2-[[(1S)-1-Carboxybutyl]amino]-1-oxopropyl]octahydro-1H-indole-2-carboxylic acid
Empirical Formula (Hill Notation):
C17H28N2O5 · xH2O
CAS Number:
Molecular Weight:
340.41 (anhydrous basis)

Products

Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade