Skip to Content
Merck
Pentyl acetate puriss. p.a., ≥98.5% (GC)

Pentyl acetate

Pentyl acetate puriss. p.a., ≥98.5% (GC)
Synonym(s):
Amyl acetate
Linear Formula:
CH3COO(CH2)4CH3
CAS Number:
Molecular Weight:
130.18
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing