Skip to Content
Merck
Penconazol PESTANAL®, analytical standard

Penconazol

Penconazol PESTANAL®, analytical standard
Empirical Formula (Hill Notation):
C13H15Cl2N3
CAS Number:
Molecular Weight:
284.18
EC Number:

Products

Shipping
Product Category
Brand
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing