Skip to Content
Merck
o-Toluenesulfonamide United States Pharmacopeia (USP) Reference Standard

o-Toluenesulfonamide

o-Toluenesulfonamide United States Pharmacopeia (USP) Reference Standard
Synonym(s):
o-Toluenesulfonamide
Linear Formula:
CH3C6H4SO2NH2
CAS Number:
Molecular Weight:
171.22

Products

Product Category
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Purity
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Package Size
API Family
Reference Material Type
Special Grade
Technique