![Nitrendipine European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/398/512/959f6d12-f354-41a3-a8f3-15de0f1be7bb/640/959f6d12-f354-41a3-a8f3-15de0f1be7bb.png)
Nitrendipine
![Nitrendipine European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/398/512/959f6d12-f354-41a3-a8f3-15de0f1be7bb/640/959f6d12-f354-41a3-a8f3-15de0f1be7bb.png)
Synonym(s):
1,4-Dihydro-2,6-dimethyl-4-(3-nitrophenyl)-3,5-pyridinedicarboxylic acid ethyl methyl ester
Empirical Formula (Hill Notation):
C18H20N2O6
CAS Number:
Molecular Weight:
360.36
Products
Product Category
Brand
Color
Formula Weight
Gene Alias
Manufacturer
Physical Form
Purity
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing