Skip to Content
Merck
Neutral Red Solution (0.33%) 3.3 g/L in DPBS, sterile-filtered, BioReagent, suitable for cell culture

Neutral Red Solution (0.33%)

Neutral Red Solution (0.33%) 3.3 g/L in DPBS, sterile-filtered, BioReagent, suitable for cell culture
Synonym(s):
Neutral Red solution, 3-Amino-7-dimethylamino-2-methylphenazine hydrochloride, Toluylene red
Empirical Formula (Hill Notation):
C15H17ClN4
CAS Number:
Molecular Weight:
288.78

Products

Brand
Formula Weight
Physical Form
Product Line
Sterilization Method
Technique
Showing 1-1 of 1