
Nervonic acid

Synonym(s):
cis-15-Tetracosenoic acid, Selacholeic acid
Linear Formula:
CH3(CH2)7CH=CH(CH2)13COOH (cis)
CAS Number:
Molecular Weight:
366.62
Products
Product Category
Brand
Biological Source
Formula Weight
Melting Point (°C)
Physical Form
Purity
Format
Quality Segment
Functional Group
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing