Skip to Content
Merck
vibrant-m

Moxonidine

vibrant-m
Synonym(s):
4-Chloro-N-(4,5-dihydro-1H-imidazol-2-yl)-6-methoxy-2-methyl-5-pyrimidinamine
Empirical Formula (Hill Notation):
C9H12ClN5O
CAS Number:
Molecular Weight:
241.68

Products

Formula Weight
Manufacturer
Physical Form
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2