
Monoclonal Anti-CDKL5 antibody produced in mouse

Synonym(s):
CFAP247, EIEE2, STK9, Anti-STK9, Anti-cyclin-dependent kinase-like 5
Products
Product Category
Brand
Biological Source
Clonality
Gene Alias
Isotype
Physical Form
Product Line
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing