Monoclonal Anti-β-Tubulin antibody produced in mouse
Synonym(s):
Monoclonal Anti-β-Tubulin
Products
Product Category
Brand
Biological Source
Clonality
Feature
Gene Alias
Isotype
Antibody Form
Research Area
Conjugate
Quality Segment
Application
Post Translational Modification
Species Reactivity
Technique
Showing 1-2 of 2