Skip to Content
Merck
Moexipril Related Compound F pharmaceutical secondary standard, certified reference material

Moexipril Related Compound F

Moexipril Related Compound F pharmaceutical secondary standard, certified reference material
Synonym(s):
N-[(S)-(+)-1-(Ethoxycarbonyl)-3-phenylpropyl]-L-alanine
Linear Formula:
C6H5CH2CH2CH(CO2C2H5)NHCH(CH3)CO2H
CAS Number:
Molecular Weight:
279.33

Products

Brand
Formula Weight
Manufacturer
Melting Point (°C)
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Package Size
API Family
Reference Material Type
Special Grade