
Millicell® Standing Cell Culture Inserts

Products
Product Category
Brand
Color
Binding Type
Diameter (mm)
Height (mm)
Surface Area cm²
Wells
Working Volume (ml)
Detection Method
Quality Segment
Hydrophobicity
Application
Color
Housing Material
Membrane Material
Package Size
Pore size (µm)
Sterility
Sterilization Method
Technique
Showing 1-10 of 10
Compare
Product Number
Description
Pricing