Skip to Content
Merck
Methyl 3-hydroxyhexanoate analytical standard

Methyl 3-hydroxyhexanoate

Methyl 3-hydroxyhexanoate analytical standard
Linear Formula:
CH3CH2CH2CH(OH)CH2CO2CH3
CAS Number:
Molecular Weight:
146.18
EC Number:

Products

Product Category
Brand
Biological Source
Formula Weight
Purity
Agency
Format
Quality Segment
Application
Organoleptic Class
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing