![Mescaline hydrochloride solution 1 mg/mL in methanol ((as free base)), certified reference material, ampule of 1 mL, Cerilliant®](/deepweb/assets/sigmaaldrich/product/structures/421/542/91c9ca3d-25a0-4927-b01e-de7375bca5de/640/91c9ca3d-25a0-4927-b01e-de7375bca5de.png)
Mescaline hydrochloride solution
![Mescaline hydrochloride solution 1 mg/mL in methanol ((as free base)), certified reference material, ampule of 1 mL, Cerilliant®](/deepweb/assets/sigmaaldrich/product/structures/421/542/91c9ca3d-25a0-4927-b01e-de7375bca5de/640/91c9ca3d-25a0-4927-b01e-de7375bca5de.png)
Synonym(s):
Mescaline hydrochloride solution
Empirical Formula (Hill Notation):
C11H17NO3·HCl
CAS Number:
Molecular Weight:
247.72
Products
Brand
Formula Weight
Manufacturer
Format
Quality Segment
Package Size
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing