Skip to Content
Merck
Medroxyprogesterone 17-acetate VETRANAL®, analytical standard

Medroxyprogesterone 17-acetate

Medroxyprogesterone 17-acetate VETRANAL®, analytical standard
Synonym(s):
17α-Acetoxy-6α-methylprogesterone, 17α-Hydroxy-6α-methyl-4-pregnene-3,20-dione 17-acetate, 6α-Methyl-17α-acetoxyprogesterone, 6α-Methyl-17α-hydroxyprogesterone acetate
Empirical Formula (Hill Notation):
C24H34O4
CAS Number:
Molecular Weight:
386.52
EC Number:

Products

Shipping
Product Category
Brand
Biological Source
Formula Weight
Gene Alias
Melting Point (°C)
Physical Form
Product Line
Purity
Format
Research Area
Quality Segment
Application
Reference Material Type
Special Grade
Sterility
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing