![Maltitol European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/128/724/a2cd299d-04bc-4ff6-8fbb-8864c38dc04e/640/a2cd299d-04bc-4ff6-8fbb-8864c38dc04e.png)
Maltitol
![Maltitol European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/128/724/a2cd299d-04bc-4ff6-8fbb-8864c38dc04e/640/a2cd299d-04bc-4ff6-8fbb-8864c38dc04e.png)
Synonym(s):
4-O-α-Glucopyranosyl-D-sorbitol
Empirical Formula (Hill Notation):
C12H24O11
CAS Number:
Molecular Weight:
344.31
Products
Product Category
Brand
Color
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Purity
Agency
Format
Analyte Suitability
Quality Segment
Application
Optical Isomer
API Family
Reference Material Type
Special Grade
Technique