Skip to Content
Merck
vibrant-m

Lynestrenol

vibrant-m
Synonym(s):
(17α)-19-Norpregn-4-en-20-yn-17-ol
Empirical Formula (Hill Notation):
C20H28O
CAS Number:
Molecular Weight:
284.44

Products

Formula Weight
Manufacturer
Physical Form
Format
Application
API Family
Reference Material Type
Special Grade