Skip to Content
Merck
L-Threonine-1-13C endotoxin tested, 99 atom % 13C, 97% (CP)

L-Threonine-1-13C

L-Threonine-1-13C endotoxin tested, 99 atom % 13C, 97% (CP)
Synonym(s):
(2S,3R)-2-Amino-3-hydroxybutyric acid-1-13C
Linear Formula:
CH3CH(OH)CH(NH2)13CO2H
CAS Number:
Molecular Weight:
120.11

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Isotope
Markush Class
Markush Group
Optical Isomer
Technique
Showing 1-2 of 2